Skip to Content
Merck
All Photos(1)

Key Documents

PZ0236

Sigma-Aldrich

CP-863187

≥98% (HPLC)

Synonym(s):

4-(2,5-Difluorophenyl)-5-(3-isopropyl-[1,2,4]triazolo[4,3-a]pyridin-6-yl)oxazole, 6-[4-(2,5-Difluorophenyl)-5-oxazolyl]-3-(1-methylethyl)-1,2,4-triazolo[4,3-a]pyridine, 6-[4-(2,5-Difluorophenyl)oxazol-5-yl]-3-isopropyl-[1,2,4]triazolo[4,3-a]pyridine, CP863187

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€108.00

€108.00


Estimated to ship on09 April 2025



Select a Size

Change View
5 MG
€108.00

About This Item

Empirical Formula (Hill Notation):
C18H14F2N4O
CAS Number:
Molecular Weight:
340.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€108.00


Estimated to ship on09 April 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

room temp

SMILES string

FC1=C(C2=C(OC=N2)C(C=C3)=CN4C3=NN=C4C(C)C)C=C(F)C=C1

InChI

1S/C18H14F2N4O/c1-10(2)18-23-22-15-6-3-11(8-24(15)18)17-16(21-9-25-17)13-7-12(19)4-5-14(13)20/h3-10H,1-2H3

InChI key

UACOXBQBNWQWOK-UHFFFAOYSA-N

Biochem/physiol Actions

CP-863187 is a potent p38α (p38α) inhibitor (IC50 = 5.8 nM) with 44-fold selectivity over p38β (p38β) and no inhibitory potency against p38γ (p38γ) and p38δ (p38δ) isoforms. CP-863187 effectively inhibits LPS-stimulated cellular TNF-α production in vitro (IC50 = 259 and 25 nM, using human whole blood and isolated mononuclear cells, respectively) and in rats in vivo (ED50 = 0.3 mg/kg p.o.) with good pharmacokinetic profile and oral availability (F = 87%/dog, 40%/monkey, 30%/rat with 5 mg/kg oral dosage; F = 66%/rat with 50 mg/kg oral dosage).
CP-863187 is a potent p38α (p38α) inhibitor.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service