Skip to Content
Merck
All Photos(1)

Key Documents

PZ0222

Sigma-Aldrich

PF-04445597

≥98% (HPLC)

Synonym(s):

1-[(1S)-1-[2-[[[[3,5-Bis(trifluoromethyl)phenyl]methyl](2-methyl-2H-tetrazol-5-yl)amino]methyl]-4-(trifluoromethyl)phenyl]-2-methylpropyl]-4-piperidinecarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€68.40
25 MG
€260.00

€68.40


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€68.40
25 MG
€260.00

About This Item

Empirical Formula (Hill Notation):
C29H31F9N6O2
CAS Number:
Molecular Weight:
666.58
UNSPSC Code:
12352200
NACRES:
NA.77

€68.40


Estimated to ship on22 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

PF-04445597 is a potent, orally bioavailable inhibitor of cholesteryl ester transfer protein (CETP).
PF-04445597 is an inhibitor of CETP.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service