Skip to Content
Merck
All Photos(1)

Key Documents

PZ0218

Sigma-Aldrich

PF-03814735

≥98% (HPLC)

Synonym(s):

N-[2-[(1S,4R)-6-[[4-(Cyclobutylamino)-5-(trifluoromethyl)-2-pyrimidinyl]amino]-1,2,3,4-tetrahydronaphthalen-1,4-imin-9-yl]-2-oxoethyl]-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25F3N6O2
CAS Number:
Molecular Weight:
474.48
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

FC(F)(F)c1c(nc(nc1)Nc3cc4c(cc3)[C@H]5N([C@@H]4CC5)C(=O)CNC(=O)C)NC2CCC2

InChI

1S/C23H25F3N6O2/c1-12(33)27-11-20(34)32-18-7-8-19(32)16-9-14(5-6-15(16)18)30-22-28-10-17(23(24,25)26)21(31-22)29-13-3-2-4-13/h5-6,9-10,13,18-19H,2-4,7-8,11H2,1H3,(H,27,33)(H2,28,29,30,31)/t18-,19+/m0/s1

InChI key

RYYNGWLOYLRZLK-RBUKOAKNSA-N

Biochem/physiol Actions

PF-03814735 is a potent inhibitor of Aurora kinase A and B (IC50 = 5 and 0.8 nM, respectively). PF-03814735 inhibits proliferation of tumor cells in culture and in xenograft tumor models. The compound PF-03814735 blocks phosphorylation of Aurora B and Histone H3, and leads to the formation of polyploid cells in culture (HCT116 and MDA-MB-231).
PF-03814735 is a potent inhibitor of Aurora kinase A and B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Paolo Fiorentino et al.
International journal of molecular sciences, 21(24) (2020-12-20)
Small cell lung cancer (SCLC) is an aggressive type of lung cancer with high mortality that is caused by frequent relapses and acquired resistance. Despite that several target-based approaches with potential therapeutic impact on SCLC have been identified, numerous targeted
Mehdi Bouhaddou et al.
Cell, 182(3), 685-712 (2020-07-10)
The causative agent of the coronavirus disease 2019 (COVID-19) pandemic, severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has infected millions and killed hundreds of thousands of people worldwide, highlighting an urgent need to develop antiviral therapies. Here we present a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service