Skip to Content
Merck
All Photos(1)

Documents

PZ0211

Sigma-Aldrich

PF-4778574

≥98% (HPLC)

Synonym(s):

N-[(3R,4S)-3-[4-(5-Cyano-2-thienyl)phenyl]tetrahydro-2H-pyran-4-yl]-2-propanesulfonamide, N-{(3R,4S)-3-[4-(5-Cyano-2-thienyl)phenyl]tetrahydro-2H-pyran-4-yl}propane-2-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N2O3S2
CAS Number:
Molecular Weight:
390.52
UNSPSC Code:
41106609
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

InChI

1S/C19H22N2O3S2/c1-13(2)26(22,23)21-18-9-10-24-12-17(18)14-3-5-15(6-4-14)19-8-7-16(11-20)25-19/h3-8,13,17-18,21H,9-10,12H2,1-2H3/t17-,18+/m1/s1

InChI key

FFAGHPLLBXWCSF-MSOLQXFVSA-N

Application

PF-4778574 may be used as a potentiator of the α-amino-3-hydroxy-5-methyl-4-isoxazole propionic acid receptor (AMPAR) in rat cortical neurons.

Biochem/physiol Actions

PF-4778574 is a potent AMPA receptor positive allosteric modulator (PAM) that has been shown to enhance cognition in animal models.

Features and Benefits

This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher L Shaffer et al.
Neuropharmacology, 153, 73-81 (2019-04-25)
Ketamine is a rapid-onset antidepressant whose efficacy long outlasts its pharmacokinetics. Multiple studies suggest ketamine's antidepressant effects require increased α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor (AMPAR)-dependent currents, which have recently been exclusively attributed to its N-methyl-d-aspartate receptor-inactive metabolite (2R,6R)-hydroxynorketamine ((2R,6R)-HNK). To investigate this
Renad Jabarin et al.
Translational psychiatry, 11(1), 234-234 (2021-04-24)
In this study we tested the hypothesis that pharmacological modulation of glutamatergic neurotransmission could rescue behavioral deficits exhibited by mice carrying a specific mutation in the Iqsec2 gene. The IQSEC2 protein plays a key role in glutamatergic synapses and mutations

Articles

We offers many products related to ion channel family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service