PZ0140
EBPC
≥98% (HPLC)
Synonym(s):
2,5-Dihydro-4-hydroxy-5-oxo-1-(phenylmethyl)-1H-pyrrole-3-carboxylic acid ethyl ester, CP-10668
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
storage condition
protect from light
color
white
solubility
DMSO: ≥10 mg/mL
storage temp.
room temp
SMILES string
CCOC(=O)C1=C(O)C(=O)N(Cc2ccccc2)C1
InChI
1S/C14H15NO4/c1-2-19-14(18)11-9-15(13(17)12(11)16)8-10-6-4-3-5-7-10/h3-7,16H,2,8-9H2,1H3
InChI key
IGYRPDIWSYGHMY-UHFFFAOYSA-N
Biochem/physiol Actions
Other Notes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Discover Bioactive Small Molecules for Lipid Signaling Research
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service