Skip to Content
Merck
All Photos(1)

Key Documents

PZ0132

Sigma-Aldrich

SC-51322

≥98% (HPLC)

Synonym(s):

8-Chloro-dibenz[b,f][1,4]oxazepine-10(11H)-carboxylic acid 2-[3-[(2-furanylmethyl)thio]-1-oxopropyl]hydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€96.60
25 MG
€300.00

€96.60


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€96.60
25 MG
€300.00

About This Item

Empirical Formula (Hill Notation):
C22H20ClN3O4S
CAS Number:
Molecular Weight:
457.93
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€96.60


Estimated to ship on22 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥10 mg/mL

storage temp.

room temp

SMILES string

Clc1ccc2Oc3ccccc3CN(C(=O)NNC(=O)CCSCc4ccco4)c2c1

InChI

1S/C22H20ClN3O4S/c23-16-7-8-20-18(12-16)26(13-15-4-1-2-6-19(15)30-20)22(28)25-24-21(27)9-11-31-14-17-5-3-10-29-17/h1-8,10,12H,9,11,13-14H2,(H,24,27)(H,25,28)

InChI key

CQBVTZDISUKDSX-UHFFFAOYSA-N

Biochem/physiol Actions

SC-51322 is a selective EP1 prostanoid receptor antagonist.

Features and Benefits

This compound is featured on the Prostanoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to prostanoid receptors for your research needs.

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service