PLA0301
Goat anti-BRG1/SMARCA4 Antibody, Affinity Purified
Powered by Bethyl Laboratories, Inc.
Synonym(s):
ATP-dependent helicase SMARCA4, BAF190, BAF190A, BRG1, BRG1-associated factor 190A, BRM/SWI2-related gene 1, CSS4, FLJ39786, MRD16, RTPS2, SNF2, SNF2-BETA, SNF2-like 4, SNF2L4, SNF2LB, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4, SWI2, brahma protein-like 1, global transcription activator homologous sequence, hSNF2b, homeotic gene regulator, mitotic growth and transcription activator, nuclear protein GRB1, protein BRG-1, protein brahma homolog 1, sucrose nonfermenting-like 4
Select a Size
€354.00
Estimated to ship on14 May 2025
Select a Size
About This Item
€354.00
Estimated to ship on14 May 2025
Recommended Products
biological source
goat
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
grade
Powered by Bethyl Laboratories, Inc.
species reactivity
mouse, human
concentration
1 mg/mL
technique(s)
immunohistochemistry: 1:500-1:2,000
immunoprecipitation (IP): 2- 10 μg/mg
western blot: 1:1,000- 1:5,000
accession no.
NP_003063.2
shipped in
wet ice
storage temp.
2-8°C
Gene Information
goat ... BRG1/SMARCA4(6597)
Immunogen
Physical form
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service