Skip to Content
Merck

Skip To

PLA0299

Goat anti-RPL26 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

DBA11, L26

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 473,00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1

€ 473,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

western blot: 1:1,000- 1:5,000

accession no.

NP_000978.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

goat ... RPL26(6154)

Immunogen

The epitope recognized by PLA0299 maps to a region between residue 95 and 145 of human Ribosomal Protein L26 using the numbering given in entry NP_000978.1 (GeneID 6154).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

RPL26 is a component of the 60S ribosomal subunit. Ribosomes are composed of a small 40S subunit and a large 60S subunit that function to catalyze protein synthesis. RPL26 is a cytoplasmic protein that functions at the translational level to induce protein expression. RPL26 has been demonstrated to bind the 5′ UTR of p53 mRNA in response to DNA damage. This interaction is described as one of many mechanisms that serve to increase p53 protein levels after DNA damage.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PLA0298PLA0302PLA0307
biological source

goat

biological source

goat

biological source

goat

biological source

goat

species reactivity

human, mouse

species reactivity

human

species reactivity

human, mouse

species reactivity

mouse, human

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

clone

polyclonal

clone

polyclonal

clone

-

clone

polyclonal

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters