Skip to Content
Merck
All Photos(1)

Key Documents

PLA0283

Sigma-Aldrich

Rabbit anti-TSG101 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ESCRT-I complex subunit TSG101, TSG10, VPS23, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€354.00

€354.00


Check Cart for Availability


Select a Size

Change View
100 μL
€354.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€354.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

mouse, human

technique(s)

western blot: 1:2,000-1:10,000

accession no.

NP_006283.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... TSG101(7251)

Immunogen

The epitope recognized by PLA0283 maps to a region between residue 300 and 350 of human Tumor Susceptibility Gene 101 using the numbering given in entry NP_006283.1 (GeneID 7251).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Tumor susceptibility gene 101 protein (TSG101) is a component of the ESCRT-I complex, a regulator of vesicular trafficking process. TSG101 mediates the association between the ESCRT-0 and ESCRT-I complex. TSG101 binds to ubiquitinated cargo proteins and is required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies (MVBs). TSG101, along with CEP55, is required for completion of cytokinesis [taken from the Universal Protein Resource (UniProt) www.uniprot.org/uniprot/Q99816].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service