Skip to Content
Merck
All Photos(2)

Documents

PLA0281

Sigma-Aldrich

Rabbit anti-CDK9 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

C-2k, CDC2-related kinase, CDC2L4, CTK1, PITALRE, Serine/threonine-protein kinase PITALRE, TAK, cell division cycle 2-like protein kinase 4, cell division protein kinase 9, cyclin-dependent kinase 9 (CDC2-related kinase), serine/threonine protein kinase PITALRE, tat-associated kinase complex catalytic subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:100-1:500
immunoprecipitation (IP): 2- 10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_001252.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... CDK9(1025)

Related Categories

Immunogen

The epitope recognized by PLA0281 maps to a region between residue 322 and 372 of human Cyclin-Dependent Kinase 9 using the numbering given in entry NP_001252.1 (GeneID 1025).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Cyclin-dependent kinase 9 (CDK9) is a member of the CDK family of kinases and a component of the multiprotein complex TAK/P-TEFb, which is an elongation factor for RNA polymerase II-directed transcription and functions by phosphorylating the C-terminal domain of the largest subunit of RNA polymerase II. CDK9 forms a complex with and is regulated by its regulatory subunit cyclin T or cyclin K.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service