Skip to Content
Merck
All Photos(2)

Key Documents

PLA0280

Sigma-Aldrich

Rabbit anti-TTI1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

KIAA0406, Protein SMG10, TEL2-interacting protein 1 homolog, Tel two interacting protein 1, Tel2 interacting protein 1 homolog (S. pombe), nonsense mediated mRNA decay factor, nonsense mediated mRNA decay factor (C. elegans), smg-10, smg-10 homolog

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€354.00

€354.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€354.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€354.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_055472.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... TTI1(9675)

Immunogen

The epitope recognized by PLA0280 maps to a region between residue 1 and 50 of human Tel2 Interacting Protein 1 using the numbering given in entry NP_055472.1 (GeneID 9675).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

TEL2-interacting protein 1 homolog (TTI1) promotes assembly, stabilizes, and maintains the activity of mTORC1 and mTORC2 complexes, which regulate cell growth and survival in response to nutrient and hormonal signals [taken from the Universal Protein Resource (UniProt) www.uniprot.org/uniprot/ O43156].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service