Skip to Content
Merck
All Photos(3)

Key Documents

PLA0172

Sigma-Aldrich

Rabbit anti-PML Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

MYL, PML/RARA fusion, PP8675, RING finger protein 71, RNF71, TRIM19, inducer of, probable transcription factor PML, promyelocytic leukemia, promyelocytic leukemia protein, tripartite motif protein TRIM19, tripartite motif-containing protein 19

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€472.00

€472.00


Check Cart for Availability


Select a Size

Change View
100 μL
€472.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€472.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:1,000
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_150241.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... PML(5371)

Immunogen

The epitope recognized by PLA0172 maps to a region between residue 375 and 425 of promyelocytic leukemia using the numbering given in entry NP_150241.2 (GeneID 5371).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

PML is a member of RBCC/TRIM family of proteins that possess a RING finger domain, B-box, and coiled-coil domain. PML is a multifunctional protein involved in activities such as tumor growth suppression, hematopoietic differentiation, gene transcription, apoptotic induction, and viral immunity. PML is often involved in chromosomal translocations with the retinoic acid receptor alpha gene. PML-retinoic acid receptor translocations are associated with acute promyelocytic leukemia.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interplay between PML NBs and HIRA for H3.3 dynamics following type I interferon stimulus.
Kleijwegt, et al.
eLife, 12 (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service