Skip to Content

PLA0168

Rabbit anti-Filamin A Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ABP-280, ABPX, Actin-binding protein 280, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD, actin binding protein 280, alpha, alpha (actin binding protein 280), alpha filamin, alpha-filamin, endothelial actin-binding protein, filamin A, filamin-1, non-muscle filamin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:500- 1:2,000, immunoprecipitation (IP): 2-5 μg/mg, western blot: 1:2,000- 1:10,000

accession no.

NP_001447.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FLNA(2316)

Immunogen

The epitope recognized by PLA0168 maps to a region between residue 2597 and 2647 of human Filamin A using the numbering given in entry NP_001447.1 (GeneID 2316).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Filamin A is an actin-binding protein that crosslinks and promotes orthogonal branching of actin filaments and also functions to anchor transmembrane proteins and intracellular signaling proteins to the actin cytoskeleton. As a scaffolding protein, filamin A is able act as a link between cell membrane receptors and intracellular signaling proteins. Defects in Filamin A are associated with multiple human diseases characterized by abnormalities in bone and neurological development.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library