Skip to Content
Merck

Skip To

PLA0080

Rabbit anti-PCNA Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ATLD2, DNA polymerase delta auxiliary protein, PCNA, cyclin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 447,00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
2

€ 447,00


Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:2,000-1:10,000, immunoprecipitation (IP): 2- 5 μg/mg, western blot: 1:5,000- 1:20,000

accession no.

P12004

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... PCNA(5111)

Immunogen

The epitope recognized by PLA0080 maps to a region between residue 225 and the C-terminus (residue 261) of human proliferating cell nuclear antigen using the numbering given in SwissProt entry P12004 (GeneID 5111).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

PCNA (Proliferating Cell Nuclear Antigen) was first identified as a factor expressed in the nuclei of replicating cells. PCNA plays a role in both DNA synthesis and repair. PCNA functions as a processivity factor for DNA polymerase by acting as a DNA clamp that tethers the polymerase catalytic units to the DNA template. The structure of PCNA is that of a trimeric ring that encircles and slides along replicating DNA. PCNA interacts with and is regulated by several proteins involved in cell cycle control and check point processes.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PLA0079PLA0156PLA0150
description

Powered by Bethyl Laboratories, Inc.

description

Powered by Bethyl Laboratories, Inc.

description

Powered by Bethyl Laboratories, Inc.

description

Powered by Bethyl Laboratories, Inc.

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

species reactivity

mouse, human

species reactivity

human, mouse

species reactivity

mouse, human

species reactivity

human

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

technique(s)

western blot: 1:5,000- 1:20,000, immunoprecipitation (IP): 2- 5 μg/mg, immunohistochemistry: 1:2,000-1:10,000

technique(s)

immunoprecipitation (IP): 2- 5 μg/mg, western blot: 1:1,000- 1:10,000, immunohistochemistry: 1:2,000- 1:10,000, flow cytometry: 0.03 μg

technique(s)

immunohistochemistry: 1:200-1:1,000, western blot: 1:2,000- 1:10,000, immunoprecipitation (IP): 2-5 μg/mg

technique(s)

immunohistochemistry: 1:2,000- 1:10,000, western blot: 1:1,000-1:10,000, immunoprecipitation (IP): 2- 5 μg/mg


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

77-92-9

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters