Skip to Content
Merck
All Photos(1)

Key Documents

PLA0005

Sigma-Aldrich

Goat anti-HP1-beta Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1:1,000- 1:5,000

accession no.

NP_006798.1

shipped in

wet ice

storage temp.

2-8°C

Gene Information

Immunogen

The epitope recognized by PLA0005 maps to a region between residue 50 and 100 of human Heterochromatin Protein p25 beta (Chromobox Homolog 1) using the numbering given in entry NP_006798.1 (GeneID 10951).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Heterochromatin protein 1 beta (HP1-beta) is a heterochromatin-associated protein that bears an evolutionarily conserved chromodomain (CD) and chromoshadow domain (CSD) that mediates chromatin and protein-protein interactions, respectively. HP1 associates with pericentromeric DNA and plays a role in gene silencing and heterochromatin formation.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service