Skip to Content
Merck
All Photos(3)

Key Documents

PIC0006

Sigma-Aldrich

Protease Inhibitor Cocktail

ReadyShield® ready-to-use solution, for the inhibition of serine, cysteine, aspartic proteases and aminopeptidases, for use in tissue culture media, Non-freezing solution

Synonym(s):

Protease inhibitor cocktail

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€156.00
5 ML
€517.00

€156.00


Check Cart for Availability


Select a Size

Change View
1 ML
€156.00
5 ML
€517.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.28
form:
ready-to-use solution
solubility:
water: soluble
storage temp.:
−20°C

€156.00


Check Cart for Availability

Product Name

ReadyShield® Protease Inhibitor Cocktail, for use in tissue culture media, Non-freezing solution

description

Tissue Culture Media

form

ready-to-use solution

quality

200

packaging

pkg of 1 mL (in amber polypropylene vial)
pkg of 5 mL (in amber polypropylene vial)

solubility

water: soluble

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is a mixture of protease inhibitors with a broad specificity for the inhibition of serine, cysteine, aspartic proteases and aminopeptidases. It is designed to be used as a supplement to tissue culture medium for the prevention of secreted proteins degradation. The product has been found to be non-toxic after a 48 hours exposure to the following adherent cell lines: A431, CHO, COS, HepG2, and HeLa, and to following non-adherent cell lines: Jurkat and HL-60. This cocktail is supplied as a ready-to-use solution using a proprietary non-freezing formulation. ReadyShield PIC0006 is a non-freezing formulation that contains the same inhibitors as is the DMSO based cocktail P1860.

Features and Benefits

  • Does not freeze in storage for quick, convenient use.
  • Inhibits serine, cysteine, aspartic proteases and aminopeptidases.
  • Tailored for use in tissue culture media.
  • Suggested as 100 X solution.
  • Reclosable screw-top vials.
  • Can be stored in cooler for approx. 1 month or room temperature for a week.

Preparation Note

It is recommended to determine the appropriate dilution for each specific cell line. This testing should begin with a dilution of at least a 200-fold to prevent deleterious effect of the solvent to cell growth.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

ReadyShield is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ReadyShield® phosphatase and protease inhibitor cocktail FAQ for sample protection in a variety of cell types and tissue extracts, including mammalian, plant, and microbial samples. Our ReadyShield® Protease Inhibitor Cocktail is a non-freezing solution that contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service