Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Check Cart for Availability | € 1.600,00 |
About This Item
Quality Segment
packaging
pkg of 1 plates
manufacturer/tradename
IROA Technologies
drug control
Új pszichoaktív anyag / New psychoactive substance (Hungary), 78/2022. (XII. 28.) BM rendelet
plate size
96 wells
shipped in
dry ice
storage temp.
−20°C
General description
Application
Features and Benefits
Preparation Note
Other Notes
To download the software package, visit sigma.com/mlsdiscovery.
To view the MLSDiscovery Video visit: MLSDiscovery Tutorial Video.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename IROA Technologies | manufacturer/tradename IROA Technologies | manufacturer/tradename IROA Technologies | manufacturer/tradename IROA Technologies |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| packaging pkg of 1 plates | packaging pkg of 1 plates | packaging pkg of 1 plates | packaging pkg of 1 plates |
| plate size 96 wells | plate size 96 wells | plate size 96 wells | plate size 96 wells |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| drug control Új pszichoaktív anyag / New psychoactive substance (Hungary), 78/2022. (XII. 28.) BM rendelet | drug control Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet | drug control - | drug control - |
signalword
Danger
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 - STOT SE 3
target_organs
Central nervous system, Respiratory system
Storage Class
3 - Flammable liquids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





