Skip to Content
Merck
All Photos(2)

Key Documents

PHG0002

SAFC

TRIS hydrochloride

Synonym(s):

Trizma® hydrochloride, TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
EC Number:
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21

biological source

synthetic

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and trace metals; tested

useful pH range

7.0-9.0

pKa (25 °C)

8.1

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tris HCl is a commonly used biological buffer. Tris/Tris HCl as a buffering system has a very broad usage in biological systems for biopharmaceutical manufacturing, cell culture, diagnostics and molecular biology. The pKa of Tris is 7.77 which corresponds closely to a physiological pH of 7.36.

Tris HCl is used as the conjugate buffer component with Tris/Tromethamine/Trizma in biopharmaceutical buffer formulations (both upstream and downstream applications), electrophoresis separations and diagnostic reagent formulations.

Other Notes

The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Forte et al.
Nature communications, 12(1), 6137-6137 (2021-10-23)
The mammalian brain stores and distinguishes among episodic memories, i.e. memories formed during the personal experience, through a mechanism of pattern separation computed in the hippocampal dentate gyrus. Decision-making for food-related behaviors, such as the choice and intake of food
Tibor Maliar et al.
Chemistry & biodiversity, 12(11), 1678-1695 (2015-11-17)
The content of biogenic compounds and the biological activities of barley (Hordeum vulgare L.)-grain extracts was evaluated. The sufficiently large and heterogeneous set of barley genotypes (100 accessions) enabled the selection of special genotypes interesting for potential industrial, pharmaceutical, and
Hong Peng et al.
Autophagy, 16(4), 698-708 (2019-06-27)
SQSTM1/p62 (sequestosome 1) is a critical macroautophagy/autophagy receptor that promotes the formation and degradation of ubiquitinated aggregates. SQSTM1 can be modified by ubiquitination, and this modification modulates its autophagic activity. However, the molecular mechanisms underpinning its reversible deubiquitination have never
Yingning Zheng et al.
Analytica chimica acta, 834, 45-50 (2014-06-15)
Our present work aimed at developing a pseudo triple-enzyme cascade electrocatalytic electrochemical aptasensor for determination of thrombin with the amplification of alcohol dehydrogenase (ADH)-Pt-Pd nanowires bionanocomposite and hemin/G-quadruplex structure that simultaneously acted as NADH oxidase and HRP-mimicking DNAzyme. With the
Michiya Matsusaki et al.
Journal of biomedical materials research. Part A, 103(10), 3386-3396 (2015-04-09)
We developed a human skin equivalent (HSE) containing blood and lymph-like capillary networks using a cell coating technique, which is a rapid fabrication technology of three-dimensional cellular constructs by cell surface coating using layer-by-layer assembled nanofilms of extracellular matrices. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service