Skip to Content
Merck

PCG2016

TruPAGE Precast Gels

4-20%, 10 x 8cm, 17-well

Synonym(s):

Precast Gels for SDS-PAGE, Precast Polyacrylamide Gels

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NB.22
UNSPSC Code:
41105300
Cassette w × l:
10 cm × 8 cm
Gel thickness:
1 mm
Compatibility:
for use with Hoefer Mighty Small II and Tall Mighty Small (SE 260 and SE280)
Packaging:
box of 10 ea
Shelf life:
24 mo. at 2‑8 °C (from the date of manufacture)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

for use with Bio-Rad Mini-PROTEAN™ Tetra Cell, Mini-PROTEAN Dodeca™, Mini-PROTEAN II and 3 cells reverse device gasket to flat side

shelf life

24 mo. at 2‑8 °C (from the date of manufacture)

packaging

box of 10 ea

concentration

4-20% polyacrylamide

cassette W × L

10 cm × 8 cm

gel thickness

1 mm

well size × volume

17 wells × 17 μL

compatibility

for use with Hoefer Mighty Small II and Tall Mighty Small (SE 260 and SE280)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

TruPAGE Precast Gels 4-20% have been used to separate proteins through sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE), followed by Western blotting.

Features and Benefits

TruPAGE SDS-PAGE Precast Gels provide improved protein resolution, tear resistance, and an extended shelf-life. TruPAGE gels are formulated with Triethanolamine (TEA)-Tricine, featuring a neutral pH running environment that helps prevents protein modification during the course of the experiment.
The precast polyacrylamide gels come in two gel cassette sizes (10 x 10 cm and 10 x 8 cm) for increased equipment compatibility. TruPAGE gels have extra tall wells to prevent lane-to-lane overflow and come in two well formats (12-well and 17-well). TruPAGE gels are available in several different gel concentrations (10%, 12%, 4-8%, 4-12% and 4-20%) to provide desired resolution of proteins of any size. Please refer to the migration chart to select the appropriate precast gel and running buffer combination for your experimental needs. Only use TruPAGE formulated buffers when running TruPAGE gels to ensure optimal results and reproducibility. Please review the TruPAGE Technical Bulletin for buffer formulation recipes and general running guidelines.

  • Long Shelf Life - 2 years from the date of manufacture when stored properly.
  • High Performance - Extended full length resoling gels provides improved resolution.
  • Durable - Stronger than standard polyacrylamide gels, preventing tearing during post-electrophoresis processing.
  • Extra Tall Wells - Eliminate sample spill over and accommodate larger sample loading volumes with well teeth that protrude above the back plate of the gel cassette.
  • Excellent Compatibility - TruPAGE gels are compatible with the popular gel running equipment, including Sigma-Aldrich′s Dual Run Tank (Z741768) and other manufacturers like Life Technologies, Bio-Rad®, Lonza®, Hoefer®, CBS Scientific as well as others, allowing you to continue using the equipment you already own.
  • Fast - Run times as low as 30 minutes.
  • Great Value - Gels and buffers offer a cost-saving alternative without sacrificing performance and experimental integrity.
TruPAGE Precast Gels Migration Chart
There is no comb to remove, which prevents torn or bent wells. Instead, the cassettes lock well fingers in place. There is also no tape to remove.

Legal Information

Bio-Rad is a registered trademark of Bio-Rad Laboratories, Inc.
Hoefer is a registered trademark of Hoefer, Inc.
Lonza is a registered trademark of Lonza Group Ltd.
TruPAGE is a trademark of Sigma-Aldrich Co. LLC

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of the effects of metformin on MDA-MB-231 breast cancer cells in a monolayer culture and in tumour spheroids as a function of nutrient concentrations.
Bizjak M, et al.
bioRxiv, 351742-351742 (2018)
Differential regulation of taurine biosynthesis in rainbow trout and Japanese flounder.
Wang Z, et al.
Scientific Reports, 6, 21231-21231 (2016)

Global Trade Item Number

SKUGTIN
PCG2016-10EA04061835518395
PCG2016-2EA04061834406280

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service