Skip to Content
Merck

Skip To

P9872

Psora-4

≥98% (HPLC)

Synonym(s):

5-(4-Phenylbutoxy)psoralen

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C21H18O4
CAS Number:
Molecular Weight:
334.37
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
protect from light, under inert gas


assay

≥98% (HPLC)

form

powder

storage condition

protect from light, under inert gas

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1Oc2cc3occc3cc2C(OCCCCc4ccccc4)=C1

InChI

1S/C21H18O4/c22-20-10-9-16-19(25-20)14-18-17(11-13-23-18)21(16)24-12-5-4-8-15-6-2-1-3-7-15/h1-3,6-7,9-11,13-14H,4-5,8,12H2

InChI key

JJAWGNIQEOFURP-UHFFFAOYSA-N

Biochem/physiol Actions

Psora-4 is a potent KV1.3 potassium channel blocker.
The lymphocyte potassium channel Kv1.3 is regarded as a new target for immunosuppression. Psora-4 is the most potent small-molecule Kv1.3 blocker known. It blocked Kv1.3 in a use-dependent manner, with a Hill coefficient of 2 and an EC50 value of 3 nM, by preferentially binding to the C-type inactivated state of the channel. It exhibited 17- to 70-fold selectivity for Kv1.3 over closely related Kv1-family channels (Kv1.1, Kv1.2, Kv1.4, and Kv1.7) with the exception of Kv1.5 (EC50, 7.7 nM) and showed no effect on human ether-a-go-go-related channel, Kv3.1, the calcium-activated K+ channels (IKCa1, SK1-SK3, and BKCa), or the neuronal NaV1.2 channel. In a test of in vivo toxicity in rats, Psora-4 did not display any signs of acute toxicity after five daily subcutaneous injections at 33 mg/kg body weight. Psora-4 selectively suppressed the proliferation of human and rat myelin-specific effector memory T cells with EC50 values of 25 and 60 nM, respectively, without persistently suppressing peripheral blood naive and central memory T cells. Because autoantigen-specific effector memory T cells contribute to the pathogenesis of T cell-mediated autoimmune diseases such as multiple sclerosis, Psora-4 and other Kv1.3 blockers may be useful as immunomodulators for the therapy of autoimmune disord.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters