Skip to Content
Merck

Skip To

P9249

Monoclonal Anti-p53−Biotin antibody produced in mouse

~1 mg/mL, clone DO-1, ascites fluid, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
biotin conjugate
Clone:
DO-1, monoclonal
Application:
FACS, ICC, IHC, IP, WB
Citations:
11


biological source

mouse

conjugate

biotin conjugate

antibody form

ascites fluid

antibody product type

primary antibodies

clone

DO-1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~53 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

flow cytometry: suitable, immunocytochemistry: 5-10 μg/mL using human A431, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: 0.5-1 μg/mL using human A431 cell extracts

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TP53(7157)

General description

p53 is a tumor suppressor gene expressed in a wide variety of tissues. It is a tetrameric nuclear DNA-binding phosphoprotein. The gene encoding it is localized on human chromosome 17p13.1 and mouse chromosome 11.

Immunogen

recombinant human wild type p53. The epitope resides beween amino acids 20-25 of human p53.

Application

Monoclonal Anti-p53-Biotin antibody produced in mouse has been used for Western Blotting.

Biochem/physiol Actions

Tumor suppressor p53 has the capability to induce cell cycle arrest and has a role in DNA repair, senescence and apoptosis. It binds to Simian vacuolating virus 40 (SV40) T-antigen and human papilloma virus E6 protein. The p53 gene is mutated in various cancers, such as of the breast, ovarian, bladder, colon and lung.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Preparation Note

For continuous use, store at 2-8°C for up to one month.
For extended storage, freeze in working aliquots.
Repeated freezing and thawing, or storage in “frostfree”freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Other Notes

In order to obtain the best results using various techniques and preparations, we recommend determining optimal working dilutions by titration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P7874P5813P4235
species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

rat, mouse, human

conjugate

biotin conjugate

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

DO-1, monoclonal

clone

DO-2, monoclonal

clone

BP53-12, monoclonal

clone

polyclonal

Gene Information

human ... TP53(7157)

Gene Information

human ... TP53(7157)

Gene Information

human ... TP53(7157)

Gene Information

human ... TP53(7157)
mouse ... Trp53(22059)
rat ... Tp53(24842)

biological source

mouse

biological source

mouse

biological source

mouse

biological source

sheep

antibody form

ascites fluid

antibody form

purified from hybridoma cell culture

antibody form

ascites fluid

antibody form

fractionated antiserum


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters