Skip to Content
Merck
All Photos(1)

Key Documents

P9036

Sigma-Aldrich

Methyl pentacosanoate

≥99% (capillary GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)23COOCH3
CAS Number:
Molecular Weight:
396.69
Beilstein:
1799674
EC Number:
MDL number:
UNSPSC Code:
12352211

Assay

≥99% (capillary GC)

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C26H52O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(27)28-2/h3-25H2,1-2H3

InChI key

WOPKHAQDUMDJIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service