Skip to Content
Merck
All Photos(1)

Key Documents

P8813

Sigma-Aldrich

Protriptyline hydrochloride

≥99% (TLC), powder

Synonym(s):

N-Methyl-5H-dibenzo[a,d]cycloheptene-5-propanamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N · HCl
CAS Number:
Molecular Weight:
299.84
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99% (TLC)

form

powder

color

white to off-white

solubility

H2O: 50 mg/mL

SMILES string

Cl[H].CNCCCC1c2ccccc2C=Cc3ccccc13

InChI

1S/C19H21N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-10,12-13,19-20H,6,11,14H2,1H3;1H

InChI key

OGQDIIKRQRZXJH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Norepinephrine uptake blocker.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Shelf-life of the powder is at least three years at room temperature.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M M Makhay et al.
Pharmacology, biochemistry, and behavior, 63(2), 319-324 (1999-06-17)
The ability of indirectly acting agonists such as norepinephrine uptake inhibitors, serotonin reuptake inhibitors, and atypical antidepressants to substitute for clenbuterol, a beta-2 adrenergic agonist, was examined in rats trained to discriminate 0.03 mg/kg clenbuterol and saline using a fixed-ratio
B Wroblewski et al.
Brain injury, 7(4), 353-362 (1993-07-01)
The results of a series of eight individual case reports in which protriptyline, an activating tricyclic antidepressant, was used as a 'stimulant' medication are presented. For some patients with head injury, traditional stimulants, such as methylphenidate, or dopaminergic stimulants, such
B T Duhart et al.
Xenobiotica; the fate of foreign compounds in biological systems, 29(7), 733-746 (1999-08-24)
1. The potential of various fungi to metabolize protriptyline (an extensively used antidepressant) was studied to investigate similarities between mammalian and microbial metabolism. 2. Metabolites produced by each organism were isolated by high-pressure liquid chromatography and identified by nuclear magnetic
Y Huang
British journal of pharmacology, 117(3), 533-539 (1996-02-01)
1 The effects of noradrenaline (NA) uptake inhibitors on contractions induced by NA, high K+, and 12-O-tetradecanoylphorbol-13-acetate (TPA) in rat isolated aorta were investigated. 2 Protriptyline (0.3 microM) and amitriptyline (0.3 microM) produced an approximately parallel shift to the right
Nonsurgical management of the obstructive sleep apnea patient.
W K Thornton et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 54(9), 1103-1108 (1996-09-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service