Skip to Content
Merck
All Photos(2)

Key Documents

P8556

Sigma-Aldrich

Pentachloronitrobenzene

suitable for plant cell culture, BioReagent, ≥94% (GC), powder

Synonym(s):

PCNB, Quintozene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Cl5NO2
CAS Number:
Molecular Weight:
295.33
Beilstein:
1914324
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Assay

≥94% (GC)

form

crystalline powder
powder

technique(s)

cell culture | plant: suitable

mp

140-143 °C (lit.)

solubility

toluene: 50 mg/mL, clear, faintly to slightly yellow

density

1.718 g/cm3 at 25 °C

antibiotic activity spectrum

fungi

application(s)

agriculture

Mode of action

DNA synthesis | interferes

SMILES string

[O-][N+](=O)c1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6Cl5NO2/c7-1-2(8)4(10)6(12(13)14)5(11)3(1)9

InChI key

LKPLKUMXSAEKID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pentachloronitrobenzene (PCNB) is used as a fungicide to suppress the growth of fungi in selective culture media such as Nash and Snyder medium (NS).

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Didem Okutman Tas et al.
Journal of agricultural and food chemistry, 55(14), 5390-5398 (2007-06-08)
The effect of pH and temperature on the microbial reductive transformation of pentachloronitrobenzene (PCNB), an organochlorine fungicide, was investigated with a mixed fermentative/methanogenic culture developed from a contaminated estuarine sediment. Culture series were incubated at a temperature range from 4
Kazuhiro Takagi et al.
Applied and environmental microbiology, 75(13), 4452-4458 (2009-05-12)
A novel aerobic pentachloronitrobenzene-degrading bacterium, Nocardioides sp. strain PD653, was isolated from an enrichment culture in a soil-charcoal perfusion system. The bacterium also degraded hexachlorobenzene, a highly recalcitrant environmental pollutant, accompanying the generation of chloride ions. Liberation of (14)CO(2) from
Ying Ying Li et al.
Journal of environmental management, 119, 143-150 (2013-03-12)
Pentachloronitrobenzene (PCNB) is a fungicide belonging to the organochlorine family and used extensively in agriculture for crop production. Many studies have implied that PCNB has become an environmental concern due to its widespread contamination in eco-systems. However, whether PCNB is
Wang Zhang et al.
Journal of hazardous materials, 198, 275-281 (2011-11-15)
Pentachloronitrobenzene (PCNB) has been identified as a potential persistent organic pollutant, and received concerns from many countries for regulation due to its impurities such as dioxins and furans. Considering the need for its disposal, the mechanochemical destruction approach was investigated
Weiguo Zhang et al.
Rapid communications in mass spectrometry : RCM, 20(10), 1563-1568 (2006-04-22)
The effects of overlapping levels and concentration ratios of overlapping components, and of scan rates of the mass spectrometer, on the capability of the automated mass spectral deconvolution and identification system (AMDIS) in pesticide residue analysis were studied. To investigate

Articles

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service