Skip to Content
Merck
All Photos(1)

Key Documents

P8477

Sigma-Aldrich

Picotamide

Synonym(s):

4-Methoxy-N,N′-bis(3-pyridinylmethyl)-1,3-benzenedicarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€156.00

€156.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€156.00

About This Item

Empirical Formula (Hill Notation):
C21H20N4O3
CAS Number:
Molecular Weight:
376.41
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

€156.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥98%

form

powder

solubility

ethanol: 50 mg/mL, clear, colorless to light yellow

SMILES string

COc1ccc(cc1C(=O)NCc2cccnc2)C(=O)NCc3cccnc3

InChI

1S/C21H20N4O3/c1-28-19-7-6-17(20(26)24-13-15-4-2-8-22-11-15)10-18(19)21(27)25-14-16-5-3-9-23-12-16/h2-12H,13-14H2,1H3,(H,24,26)(H,25,27)

InChI key

KYWCWBXGRWWINE-UHFFFAOYSA-N

Gene Information

human ... TBXA2R(6915)

Biochem/physiol Actions

Eicosenoid receptor antagonist; a thromboxane A2 antagonist; antiplatelet agent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Pancera et al.
Journal of internal medicine, 242(5), 373-376 (1998-01-04)
To assess the role of thromboxane A2 and of angiotensin II in patients with primary Raynaud's phenomenon. After an eight-day run-in period, the patients were enrolled in a single-blind, cross-over, study. Patients were examined at the Ambulatory for Microcirculatory Diseases
P L Malini et al.
Lancet (London, England), 350(9070), 15-18 (1997-07-05)
The increased prostaglandin synthesis that might follow stimulation of the arachidonic acid cascade by angiotensin-converting-enzyme inhibition (ACE-I) has been suggested to underlie the appearance of cough on ACE-I treatment. We investigated whether the prostanoid thromboxane was involved. Nine patients with
M Berrettini et al.
European journal of clinical pharmacology, 39(5), 495-500 (1990-01-01)
Picotamide (G 137), a new non prostanoid inhibitor of in vitro arachidonic acid induced platelet aggregation, has been further characterized in in vitro and ex vivo studies. When whole blood was activated with collagen in the presence of picotamide 5
W Van Molle et al.
FEBS letters, 467(2-3), 201-205 (2000-02-17)
Tumor necrosis factor (TNF) induces hepatitis when injected in human beings or in rodents. The molecular mechanism by which TNF induces hepatic distress remains largely unknown, although induction of apoptosis of hepatocytes appears to be an essential step. In order
G D'Andrea et al.
Neurological sciences : official journal of the Italian Neurological Society and of the Italian Society of Clinical Neurophysiology, 27 Suppl 2, S96-S99 (2006-05-12)
Migraine with aura (MwA) sufferers, at times, need specific treatments. This is the case when the auras are frequent, prolonged and cause anxiety and distress. Abnormal release of glutamate, which may trigger auras, and abnormal platelet behaviour, which constitutes a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service