Skip to Content
Merck
All Photos(1)

Key Documents

P8299

Parafilm® "M" cutter/dispenser

acrylic

Synonym(s):

Parafilm Dispenser, Parafilm M Cutter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.35

material

acrylic

manufacturer/tradename

Bel-Art

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

Adapts to 2- or 4-inch roll. 6 in. × 6 in. × 8 in. (H × W × D)

Legal Information

Parafilm is a registered trademark of Bemis Company, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service