Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Check Cart for Availability | € 304,00 |
About This Item
NACRES:
NB.13
UNSPSC Code:
41161502
€ 304,00
Check Cart for Availability
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37861-0000
color
none
General description
The plate reader has a clear deck for resting plates and a large, clear, adjustable mirror for viewing their underside. This tool allows safe examination of the bottom of the plates without risking spills on the face and eyes. It is ideal for Hemagglutination, Hemagglutination Inhibition, Hemolysis and Agglutination testing. Other applications include counting of bacterial cell cultures for colony or plaque growth. Mirror magnification is 3X actual plate size. Polypropylene with an acrylic deck.
Features and Benefits
- Easy, magnified viewing; hands-free
- For microplates, view color changes and locate cells for further analysis
- For petri dishes, view cell growth and take advantage of hands-free design to make notes and isolate cell colonies
- Ideal for evaluating monolayer cell growth
Legal Information
Scienceware is a registered trademark of Bel-Art Products, a division of SP
1 of 1
This Item | |||
|---|---|---|---|
| packaging pack of 1 ea | packaging pkg of 1 ea | packaging pack of 25, case of 50 | packaging pack of 25, case of 50 |
| manufacturer/tradename Bel-Art F37861-0000 | manufacturer/tradename Bel-Art F37848-0000 | manufacturer/tradename Corning 3935 | manufacturer/tradename Corning 3099 |
| sterility non-sterile | sterility non-sterile | sterility sterile | sterility sterile |
| color none | color none | color black | color clear |
Choose from one of the most recent versions:
Already Own This Product?