Skip to Content
Merck
All Photos(1)

Key Documents

P7575

Sigma-Aldrich

L-trans-Pyrrolidine-2,4-dicarboxylic acid

≥98%

Synonym(s):

(2S,4R)-Pyrrolidine-2,4-Dicarboxylic Acid, L-trans-2,4-PDC, trans-4-Carboxy-L-proline

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
€60.80
10 MG
€193.00

€60.80


Estimated to ship on23 April 2025


Request a Bulk Order

Select a Size

Change View
2 MG
€60.80
10 MG
€193.00

About This Item

Empirical Formula (Hill Notation):
C6H9NO4
CAS Number:
Molecular Weight:
159.14
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

€60.80


Estimated to ship on23 April 2025


Request a Bulk Order

Quality Level

Assay

≥98%

form

powder

SMILES string

OC(=O)[C@H]1CN[C@@H](C1)C(O)=O

InChI

1S/C6H9NO4/c8-5(9)3-1-4(6(10)11)7-2-3/h3-4,7H,1-2H2,(H,8,9)(H,10,11)/t3-,4+/m1/s1

InChI key

NRSBQSJHFYZIPH-DMTCNVIQSA-N

Related Categories

Application

L-trans-Pyrrolidine-2,4-dicarboxylic acid has been used:
  • as a glutamate transport inhibitor to test its impairing effect on glutamate-induced cell death in HT22 neuronal cells[1]
  • as an analog of glutamate to test its effect on glutamate/glutamine-dependent acid resistance of E. coli and S. flexneri[2]
  • as sodium glutamate (Na+/Glu) cotransporter inhibitor in myogenic cell line C2C12[3]

Biochem/physiol Actions

L-trans-Pyrrolidine-2,4-dicarboxylic acid is a selective glutamate transporter inhibitor.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingmei Zhang et al.
CNS neuroscience & therapeutics, 30(1), e14487-e14487 (2023-10-07)
Chronic cerebral hypoperfusion-induced demyelination causes progressive white matter injury, although the pathogenic pathways are unknown. The Single Cell Portal and PanglaoDB databases were used to analyze single-cell RNA sequencing experiments to determine the pattern of EAAT3 expression in CNS cells.
H-R Weng et al.
Neuroscience, 138(4), 1351-1360 (2006-01-24)
Glutamate is a primary excitatory neurotransmitter in the mammalian CNS. Glutamate released from presynaptic neurons is cleared from the synaptic cleft passively by diffusion and actively by glutamate transporters. In this study, the role of glutamate transporters in sensory processing
Shinichirou Oono et al.
Current eye research, 34(7), 598-605 (2009-11-11)
To determine whether pyroglutamic acid (PGA) enhances the survival of retinal ganglion cells (RGCs) after optic nerve (ON) transection in vivo and RGCs in culture. The RGCs of rats were retrogradely labeled by Fluorogold (FG)-soaked sponges placed on both superior
Younglim Lee et al.
Psychopharmacology, 191(1), 55-65 (2007-01-17)
Recent evidence in clinical and preclinical studies has implicated glutamate neurotransmissions in pathophysiology of mood disorders. The regulation of amino acid neurotransmission, i.e., glutamate and gamma-aminobutyric acid (GABA) involves coordinated mechanisms of uptake and transport within a tripartite synaptic system
V J Balcar
FEBS letters, 300(3), 203-207 (1992-04-06)
Uptake of 1 microM [3H]L-glutamate by cultured 3T3 fibroblasts was strongly dependent on extracellular Na+; it was reduced by elevated concentrations of K+ (60 mM) but it was not influenced by variations in the concentration of Ca2+ (0-9.6 mM). D-

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service