Skip to Content
Merck
All Photos(1)

Key Documents

P6990

Sigma-Aldrich

1,2-Dipalmitoylphosphatidylinositol 3,4-diphosphate sodium salt

≥98%, solid

Synonym(s):

1,2-Dipalmitoyl-L-α-phosphatidyl-D-myo-inositol 3,4-bisphosphate sodium salt, 1-(1,2-Dihexadecanoylphosphatidyl)inositol-3,4-diphosphate sodium salt, Dipalmitoyl-L-α-phosphatidyl-D-myo-inositol 3,4-bisphosphate sodium salt, Phosphatidylinositol-3,4-diphosphate C-16 sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€145.00

€145.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€145.00

About This Item

Empirical Formula (Hill Notation):
C41H81O19P3 · xNa+
Molecular Weight:
970.99 (free acid basis)
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.77

€145.00


Estimated to ship on14 April 2025


Request a Bulk Order

Quality Level

Assay

≥98%

form

solid

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(O)(=O)O[C@H]1[C@H](O)[C@@H](O)[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H]1O)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C41H81O19P3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(42)55-31-33(57-35(43)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)32-56-63(53,54)60-39-36(44)37(45)40(58-61(47,48)49)41(38(39)46)59-62(50,51)52/h33,36-41,44-46H,3-32H2,1-2H3,(H,53,54)(H2,47,48,49)(H2,

InChI key

HKWJHKSHEWVOSS-OMDJCSNQSA-N

Biochem/physiol Actions

Diphosphorylated phosphatidylinositol that may be hydrolyzed by PI-specific phospholipase C to generate IP3 and DAG.

Preparation Note

The product is soluble in aqueous detergent solutions. It forms a clear solution at 8 mg/ml in 50 mM HEPES buffer, pH 7.0, containing 5 mM EDTA and 16 mg/ml sodium deoxycholate. It is also soluble in a 3:2:1 mixture of chloroform:methanol:water at 10 mg/ml and will form an opaque suspension at 1 mg/ml in phosphate-buffered saline (PBS), pH 7.2. Aqueous solutions remain active for less than 24 hours.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service