Skip to Content
Merck

Skip To

P6491

Nunc® MaxiSorp 384 well plates

384 well plate, black, polystyrene, 120ul/well, pinchbar design, 30/cs

Synonym(s):

384 multiwell plate, black muliplate, Nunc 460518, cell cultivation plate, culture plate, multiwell plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
30 ea
Check Cart for Availability
€ 518,00

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
black polystyrene , flat bottom black polystyrene wells
Size:
384 wells
Sterility:
non-sterile
Binding type:
high binding surface
Feature:
lid: no

€ 518,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

black polystyrene , flat bottom black polystyrene wells

sterility

non-sterile

feature

lid: no

packaging

case of 30 ea (internal packs of 10)

manufacturer/tradename

Nunc 460518

maximum volume

120 μL

plate L × W

128 mm × 86 mm

size

384 wells

well maximum volume

120 μm

working volume

100 μL

color

black

suitability

suitable for (colorimetric assays, sample storage; luminescence and fluorescence assays, ELISA)

binding type

high binding surface

General description

MaxiSorp plates for ELISA

  • Same footprint as a 96 well plate
  • Working volume range of 10-100 μL
  • Compatible with robotic, automated readers and liquid handling systems
  • Assay miniaturization increases productivity and reduces cost
  • C-well for optical clarity and sample recovery
  • Reduced risk of well-to-well contamination
  • Optimizes robotic access to wells
  • White and black plates minimize crosstalk
  • Rounded square wells eliminate wicking (capillary action)
  • Flat bottom for optical reading
  • Optimized for scintillation counting
  • Black 384 well plate reduces background fluorescence and minimizes backlight scatter
  • White 384 well plate maximizes reflection of signal while maintaining low background
  • Clear 384 well plate for optical clarity for readthrough applications

Legal Information

MaxiSorp is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P6116P6366P9116
manufacturer/tradename

Nunc 460518

manufacturer/tradename

Nunc 460372

manufacturer/tradename

Nunc 464718

manufacturer/tradename

Nunc 142762

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

sterile

material

black polystyrene , flat bottom black polystyrene wells

material

flat bottom white polystyrene wells

material

clear polystyrene , flat bottom clear polystyrene wells

material

flat bottom clear polystyrene wells, white polystyrene

binding type

high binding surface

binding type

(MaxiSorp surface)

binding type

(MaxiSorp surface)

binding type

-

feature

lid: no

feature

lid: no

feature

lid: no

feature

lid, polystyrene/polymer base

well maximum volume

120 μm

well maximum volume

120 μm

well maximum volume

120 μm

well maximum volume

100 μm


Still not finding the right product?

Explore all of our products under Nunc® MaxiSorp 384 well plates




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library