Skip to Content
Merck
All Photos(1)

Key Documents

P5999

Sigma-Aldrich

Monoclonal Anti-Prion Protein antibody produced in mouse

1 mg/mL, clone IPC1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-AA960666, Anti-CD230, Anti-PRIP, Anti-PrP, Anti-PrP<C>, Anti-PrPSc, Anti-Prn-i, Anti-RP23-401J24.1, Anti-Sinc

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

IPC1, monoclonal

form

buffered aqueous solution

mol wt

antigen 25-35 kDa

species reactivity

hamster, mouse, rat

concentration

1 mg/mL

technique(s)

microarray: suitable
western blot: 0.05-0.1 μg/mL using mouse brain extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Prnp(19122)
rat ... Prnp(24686)

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Prion Protein (mouse IgG1 isotype) is derived from the hybridoma IPC1 produced by the fusion of mouse myeloma cells (NSO cells) and splenocytes from PrP knock-out mice immunized with recombinant mouse PrPc.
Prion is a cell surface glycoprotein present in two isoform- PrPC (a cellular isoform) and PrPSc (a disease associated isoform). Prion Protein encodes a membrane glycosylphosphatidylinositol-anchored glycoprotein that tends to aggregate into rod-like structures. The encoded protein contains a highly unstable region of five tandem octapeptide repeats. PrPC is found in the neurons of the brain and spinal cord. The prion protein is associated with several diseases like bovine spongiform encephalopathy, Creutzfeldt-Jakob disease and fatal familial insomnia.

Immunogen

recombinant mouse PrPC.

Application

Monoclonal Anti-Prion Protein antibody produced in mouse is useful in enzyme linked immunosorbent assay (ELISA), immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Prion protein (PrP) is a natural protein synthesized within the secretory pathway and transported to the surface of the cell where it is tethered to the cell membrane by a glycosylphosphatidylinositol (GPI) anchor. The activity of PrP is not well understood; it may be involved in copper utilization, serving to buffer copper at the synaptic cleft or to mediate re-uptake of copper into the presynapse. Alternatively, bound copper may influence PrP binding characteristics; the PrP-copper complex may be crucial for synaptic homeostasis as a result of its anti-oxidant activity. Prion plaques are of three types: unicentric (single, compact core), multicentric (two or more cores and definite border), and diffuse plaques without a definite central core.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microglia and the pathogenesis of spongiform encephalopathies.
Rezaie P and Lantos PL
Brain Research Reviews, 35(1), 55-72 (2001)
The cellular prion protein (PrP(C)): its physiological function and role in disease.
Westergard L
Biochimica et Biophysica Acta, 1772(6), 629-644 (2007)
Robert Hnasko et al.
PloS one, 4(2), e4440-e4440 (2009-02-13)
Intracerebral inoculation of 263K Scrapie brain homogenate (PrPsc) with a self-assembling RADA-peptide (RADA) significantly delayed disease onset and increased hamster survival. Time of survival was dependent on the dose of RADA and pre-incubation with PrPsc prior to inoculation. RADA treatment
Transplanted neural precursors enhance host brain-derived myelin regeneration.
Einstein O, et al.
The Journal of Neuroscience, 29(50), 15694-15702 (2009)
Prion and prejudice: normal protein and the synapse.
Brown DR.
Trends in Neurosciences, 24(2), 85-90 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service