P5510
Potassium bromide
IR Grade, suitable for IR spectroscopy
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
grade
IR Grade
vapor pressure
<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)
technique(s)
IR spectroscopy: suitable
mp
734 °C (lit.)
SMILES string
[K+].[Br-]
InChI
1S/BrH.K/h1H;/q;+1/p-1
InChI key
IOLCXVTUBQKXJR-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
May be used for removal of peripheral membrane proteins.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jun Pan et al.
Organic letters, 13(18), 4974-4976 (2011-08-26)
A facile Pd-catalyzed conversion of aryl and vinyl triflates to aryl and vinyl halides (bromides and chlorides) is described. This method allows convenient access to a variety of aryl, heteroaryl, and vinyl halides in good to excellent yields and with
Mamta J Patel et al.
Journal of cellular biochemistry, 106(2), 306-316 (2009-01-07)
Bone loss due to osteoporosis or disuse such as in paraplegia or microgravity is a significant health problem. As a treatment for osteoporosis, brief exposure of intact animals or humans to low magnitude and high frequency (LMHF) mechanical loading has
Patrizia Borghetti et al.
Langmuir : the ACS journal of surfaces and colloids, 26(24), 19007-19013 (2010-11-17)
The role of potassium in the formation of synthetic eumelanin aggregates is investigated by atomic force microscopy and soft-X-ray spectroscopy. Control over the thin film granularity is achieved by using K salts, in both drop casting and electrodeposition of eumelanin
Pengfei Zhang et al.
Journal of bacteriology, 192(14), 3608-3619 (2010-05-18)
The simultaneous nutrient germination of hundreds of individual wild-type spores of three Bacillus species and a number of Bacillus subtilis strains has been measured by two new methods, and rates of release of the great majority of the large pool
Kazue Kurihara
Advances in colloid and interface science, 158(1-2), 130-138 (2010-05-11)
Brush layers of polyelectrolytes, ionized chains of poly(glutamic acid) (PLGA) and poly(lysine) (PLL), prepared by the Langmuir-Blodgett method, were characterized using the surface forces measurements at various pHs, salt concentrations and chain densities. This paper reviews the major results: (1)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service