Skip to Content
Merck
All Photos(1)

Key Documents

P5475

Sigma-Aldrich

C-CMV-24 Sequencing Primer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

grade

Molecular Biology
for molecular biology

shipped in

dry ice

storage temp.

−20°C

General description

The C-CMV-24 sequencing primer is a chemically synthesized, 24 base, single strand oligodeoxyribonucleotide designed for dsDNA or ssDNA sequencing of FLAG® fusion proteins at the N-terminus of the pFLAG-CMV expression vectors. The primer is complementary to map position nucleotides 1116-1139 of negative, non-coding DNA strand of the pFLAG-CMV-1 vector and nucleotides 1073-1096 of negative, non-coding DNA strand of the pFLAG-CMV-2 vector.

Application

The N-CMV-30 sequencing primer is a chemically synthesized, 30 base, single strand oligodeoxyribonucleotide designed for dsDNA or ssDNA sequencing of FLAG® fusion proteins at the C-terminus of the pFLAG-CMV expression vectors. The primer is complementary to map position nucleotides 818-847 of positive, coding DNA strand of the pFLAG-CMV vectors.

Components

The primer is supplied at an initial concentration of 5 uM in 1xTE buffer. The nucleotide sequence is as follows: 5′-TAT-TAG-GAC-AAG-GCT-GGT-GGG-CAC-3′

Other Notes

(5′-TAT TAG GAC AAG GCT GGT GGG CAC-3′)

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service