Skip to Content
Merck
All Photos(1)

Key Documents

P5011

Sigma-Aldrich

Poly-γ-benzyl-L-glutamate

mol wt 30,000-70,000

Synonym(s):

PBLG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

form

solid

Quality Level

mol wt

30,000-70,000

color

white

storage temp.

−20°C

InChI

1S/C12H15NO4/c13-10(12(15)16)6-7-11(14)17-8-9-4-2-1-3-5-9/h1-5,10H,6-8,13H2,(H,15,16)/t10-/m0/s1

InChI key

BGGHCRNCRWQABU-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Molecular weight based on GPC

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Electrospun Poly(γ-Benzyl-l-Glutamate) and Its Alkali-Treated Meshes: Their Water Wettability and Cell-Adhesion Potential.
Hakamada Y, Ohgushi N, et al.
Journal of Biomaterials Science. Polymer Edition (2011)
Ipek Ozcan et al.
International journal of nanomedicine, 5, 1103-1111 (2011-01-29)
Poly(γ-benzyl-L-glutamate) (PBLG) derivatives are synthetic polypeptides for preparing nanoparticles with well controlled surface properties. The aim of this paper was to investigate the biodistribution of pegylated PBLG in rats. For this purpose, nanoparticles were prepared by a nanoprecipitation method using
Rajeswari Ravichandran et al.
Biomaterials, 33(3), 846-855 (2011-11-04)
Tissue engineering and nanotechnology have enabled engineering of nanostructured materials to meet the current challenges in bone treatment owing to rising occurrence of bone diseases, accidental damages and defects. Poly(L-lactic acid)/Poly-benzyl-L-glutamate/Collagen (PLLA/PBLG/Col) scaffolds were fabricated by electrospinning and nanohydroxyapatite (n-HA)
Zeinab Serhan et al.
Chemical communications (Cambridge, England), 46(35), 6599-6601 (2010-08-20)
NAD 2D NMR spectroscopy using chiral mesophases made of poly-gamma-benzyl-l-glutamate and pyridine allowed us to evaluate, for the first time, the natural site-specific enantio-isotopomeric excesses at each methylene group of linoleic acid, a central, essential PUFA precursor of all conjugated
Ma Elisa Martínez-Barbosa et al.
Bioconjugate chemistry, 20(8), 1490-1496 (2009-08-19)
In the present work, the possibility to obtain PEGylated nanoparticles from two PBLG derivatives, PEG-b-poly(γ-benzyl L-glutamate), PBLG-PEG-60, and poly(γ-benzyl L-glutamate), PBLG-Bnz-50, by nanoprecipitation has been investigated. Particles were prepared not only from one polymer (PBLG-PEG-60 or PBLG-Bnz-50), but also from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service