Skip to Content
Merck
All Photos(1)

Key Documents

P5001

Sigma-Aldrich

Phe-Val

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O3
CAS Number:
Molecular Weight:
264.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(C)C(NC(=O)C(N)Cc1ccccc1)C(O)=O

InChI

1S/C14H20N2O3/c1-9(2)12(14(18)19)16-13(17)11(15)8-10-6-4-3-5-7-10/h3-7,9,11-12H,8,15H2,1-2H3,(H,16,17)(H,18,19)

InChI key

IEHDJWSAXBGJIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingkun Fu et al.
Journal of the American Society for Mass Spectrometry, 21(10), 1737-1752 (2010-08-14)
Gas-phase reactivity of a positively charged aromatic σ,σ-biradical (N-methyl-6,8-didehydroquinolinium) was examined toward six aliphatic amino acids and 15 dipeptides by using Fourier transform ion cyclotron resonance mass spectrometry (FT-ICR) and laser-induced acoustic desorption (LIAD). While previous studies have revealed that
Yiqun Huang et al.
Journal of the American Chemical Society, 127(11), 3973-3978 (2005-03-18)
Gas-phase reactivity of five differently substituted positively charged phenyl radicals was examined toward six amino acids by using Fourier transform ion cyclotron resonance mass spectrometry (FT-ICR). The reactivity of the radicals studied was determined by the electrophilicity of the radical
C H Görbitz
Acta crystallographica. Section C, Crystal structure communications, 56(Pt 12), 1496-1498 (2000-12-19)
One of the amino H atoms of L-phenylalanyl-L-valine, C(14)H(20)N(2)O(3), participates in a rare secondary interaction in being accepted by the aromatic ring of the phenylalanine side chain. The phenyl group is also a donor in a weak hydrogen bond to
Sen Li et al.
The Journal of organic chemistry, 74(20), 7724-7732 (2009-09-24)
Laser-induced acoustic desorption (LIAD) incorporated with Fourier transform ion cyclotron resonance mass spectrometry (FT-ICR) has been utilized to investigate phenyl radical-induced damage to dipeptides in the gas phase. On the basis of the product branching ratios measured for the reactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service