Skip to Content
Merck
All Photos(1)

Key Documents

P4904

Sigma-Aldrich

β-Acetyl-γ-O-hexadecyl-L-α-phosphatidylcholine hydrate

≥98%

Synonym(s):

1-Hexadecyl-2-acetyl-sn-glycero-3-phosphocholine, 1-O-Palmityl-2-acetyl-sn-glycero-3-phosphocholine monohydrate, 2-Acetyl-1-hexadecyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, 1-hexadecyl-2-acetyl, PAF(C16), Platelet activating factor

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€73.80
5 MG
€120.00
10 MG
€398.00
25 MG
€557.00

€73.80


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€73.80
5 MG
€120.00
10 MG
€398.00
25 MG
€557.00

About This Item

Empirical Formula (Hill Notation):
C26H54NO7P · xH2O
CAS Number:
Molecular Weight:
523.68 (anhydrous basis)
Beilstein:
3574588
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

€73.80


Estimated to ship on22 May 2025


Request a Bulk Order

Assay

≥98%

form

powder

storage temp.

−20°C

SMILES string

O.CCCCCCCCCCCCCCCCOC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(C)=O

InChI

1S/C26H54NO7P.H2O/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-21-31-23-26(34-25(2)28)24-33-35(29,30)32-22-20-27(3,4)5;/h26H,6-24H2,1-5H3;1H2/t26-;/m1./s1

InChI key

HFMSDOHEENWULV-UFTMZEDQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-Acetyl-γ-O-hexadecyl-L-α-phosphatidylcholine has been used:
  • to study its effects on phagocytic activity and production of superoxide anion in equine alveolar macrophage and blood neutrophils[1]
  • to study its protective effects against cisplatin-induced cell death in a human melanoma cell line in vitro[2]
  • to determine its effects on cervical ripening and leukocyte infiltration in rat models[3]

Biochem/physiol Actions

β-Acetyl-γ-O-hexadecyl-L-α-phosphatidylcholine or platelet-activating factor (PAF), is a phospholipid[4], and a pro-inflammatory lipid mediator.[5] It acts as a ligand for G protein-coupled platelet-activating factor (PAF) receptors and participates in several biological processes such as airway constriction, hypotension, platelet activation, and increased vascular permeability.[5] PAF is released by various cells such as platelets, neutrophils, endothelial cells, and monocytes/macrophages. The levels of PAF are low in the blood due to its rapid metabolism by PAF acetylhydrolase and low production.[6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R N Pinckard et al.
Journal of leukocyte biology, 59(2), 219-228 (1996-02-01)
Alkyl-PAF induced two components of polymorphonuclear leukocyte (PMN) priming for enhanced fMLP- and rC5a-induced superoxide anion (O2-) production. Component A priming had a shallow, linear alkyl-PAF concentration-response slope (10 pM-1 nM), and component B priming had a significantly steeper concentration-response
T Hiran et al.
Prostaglandins & other lipid mediators, 66(4), 305-315 (2002-01-12)
Platelet-activating factor (PAF) is an early product of the inflammatory environment, influencing development and resolution of inflammation. Its production is greater in neutrophils and macrophages, which predominantly synthesize 1-alkyl sn-2 acetyl glycerophosphocholine (GPC) than in nongranulocytes (B cells and endothelial
Platelet-activating factor.
S M Prescott et al.
The Journal of biological chemistry, 265(29), 17381-17384 (1990-10-15)
Damien Garrido et al.
Frontiers in veterinary science, 4, 226-226 (2018-01-13)
Lipid mediators are known to play important roles in the onset and resolution phases of the inflammatory response in mammals. The phospholipid platelet-activating factor (PAF) is a pro-inflammatory lipid mediator which participates in vascular- and innate immunity-associated processes by increasing
Katarina Mariann Jørgensen et al.
Archives of dermatological research, 302(3), 221-227 (2009-12-31)
Platelet activating factor (PAF, 1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine) is known to be present in excess in psoriatic skin, but its exact role is uncertain. In the present study we demonstrate for the first time the role of group VI PLA(2) in PAF-induced arachidonic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service