Skip to Content
Merck
All Photos(1)

Key Documents

P4901

Sigma-Aldrich

Phorbol 12-decanoate

liquid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H46O7
CAS Number:
Molecular Weight:
518.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@@]2(O)C(C=C(CO)C[C@@]3(O)C2C=C(C)C3=O)C4C(C)(C)[C@]14O

InChI

1S/C30H46O7/c1-6-7-8-9-10-11-12-13-23(32)37-26-19(3)29(35)21(24-27(4,5)30(24,26)36)15-20(17-31)16-28(34)22(29)14-18(2)25(28)33/h14-15,19,21-22,24,26,31,34-36H,6-13,16-17H2,1-5H3/t19-,21+,22-,24-,26-,28-,29-,30-/m1/s1

InChI key

LXYSVTVLQYLWKR-LKSXOGJWSA-N

Packaging

Sealed ampule.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service