Skip to Content
Merck
All Photos(1)

Key Documents

P4646

Sigma-Aldrich

Proflavine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12171500

Pricing and availability is not currently available.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelsey J Rosbach et al.
Biomedical optics express, 1(3), 911-922 (2011-01-25)
This prospective pilot study evaluates the potential of high-resolution fiber optic microscopy (HRFM) to identify lymph node metastases in breast cancer patients. 43 lymph nodes were collected from 14 consenting breast cancer patients. Proflavine dye was topically applied to lymph

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service