Skip to Content
Merck
All Photos(2)

Key Documents

P4406

Sigma-Aldrich

Papain Agarose from papaya latex

lyophilized powder, 90-150 units/mL packed gel

Sign Into View Organizational & Contract Pricing

Select a Size

25 UNITS
€316.00
50 UNITS
€577.00
100 UNITS
€988.00

€316.00


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
25 UNITS
€316.00
50 UNITS
€577.00
100 UNITS
€988.00

About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

€316.00


Estimated to ship on28 March 2025


Request a Bulk Order

biological source

papaya (latex)

Quality Level

form

lyophilized powder

specific activity

90-150 units/mL packed gel

mol wt

21 kDa

extent of labeling

3,000-5,000 units per g agarose (at pH7.0 at 30 °C.)

matrix

beaded agarose

storage temp.

−20°C

Application

Papain is used in dissecting solutions. It is used to produce Fab fragments of antibodies.[1] It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues. Papain has been used to isolate morphologically intact cortical neurons from postnatal rats. Limited papain digestion is used for structural studies of enzymes and other proteins. In addition, papain is used in red cell serology to modify the red cell surface to enhance or destroy the reactivity of many red cell antigens. Papain is useful for platelet serology studies. Papain has also been used in the enzymatic synthesis of amino acids, peptides, and other molecules. Product P4406 is agarose-linked papain and is provided as a lyophilized powder stabilized with lactose. Product P4406 has been used to digest purified anti-JMJD6 hamster monoclonal antibody into Fab fragments[2].
Used to produce Fab fragments of antibodies. Also used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.

Biochem/physiol Actions

Papain papaya latex has antifungal activity against C. albicans. It is a cysteine protease that cleaves peptide bonds of basic amino acids, leucine, or glycine.
. The pH optimum is 6.0-7.0
. Papain hydrolyzes esters and amides. Papain consists of a single polypeptide chain with three disulfide bridges and a sulfhydryl group necessary for activity of the enzyme.
A cysteine protease that cleaves peptide bonds of basic amino acids, leucine or glycine. pH optimum 6.0-7.0
Also hydrolyzes esters and amides.

Unit Definition

One unit will hydrolyze 1.0 μmole of BAEE per min at pH 6.2 at 25°C, unless otherwise indicated below.

Physical form

Lyophilized powder, stabilized with lactose

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yafei Liu et al.
Cell, 184(13), 3452-3466 (2021-06-18)
Antibodies against the receptor-binding domain (RBD) of the SARS-CoV-2 spike protein prevent SARS-CoV-2 infection. However, the effects of antibodies against other spike protein domains are largely unknown. Here, we screened a series of anti-spike monoclonal antibodies from coronavirus disease 2019
Xia Hong et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(33), 14568-14572 (2010-08-04)
JMJD6 is a Jumonji C domain-containing hydroxylase. JMJD6 binds alpha-ketoglutarate and iron and has been characterized as either a histone arginine demethylase or U2AF65 lysyl hydroxylase. Here, we describe the structures of JMJD6 with and without alpha-ketoglutarate, which revealed a
Y. Ozari et al.
Journal of the Chemical Society. Chemical Communications, 295-295 (1974)

Articles

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service