Skip to Content
Merck
All Photos(1)

Key Documents

P4270

Supelco

5α-Pregnan-17α-ol-3,20-dione

~97%

Synonym(s):

17α-Hydroxy-5α-pregnane-3,20-dione, Allopregnane-3,20-dione-17α-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H32O3
CAS Number:
Molecular Weight:
332.48
UNSPSC Code:
41116107

Assay

~97%


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edith Bellavance et al.
Journal of medicinal chemistry, 52(23), 7488-7502 (2009-09-24)
17beta-Hydroxysteroid dehydrogenase type 7 (17beta-HSD7) catalyzes the reduction of estrone (E(1)) into estradiol (E(2)) and of dihydrotestosterone (DHT) into 5alpha-androstane-3beta,17beta-diol (3beta-diol), therefore modulating the level of mitogenic estrogens and androgens in humans. By classical and parallel chemistry, we generated several

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service