Skip to Content
Merck
All Photos(1)

Key Documents

P4148

Sigma-Aldrich

1,2-Dihexanoyl-sn-glycero-3-phosphocholine

solution, 20 mg/mL in chloroform, ≥99%

Synonym(s):

(7R)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxohexyl)oxy]-3,5,9-trioxa-4-phosphapentadecan-1-aminium 4-oxide, inner salt, L-α-Dihexanoyllecithin, L-α-Phosphatidylcholine, dicaproyl, Dicaproyl-L-α-lecithin, Dihexanoyllecithin, PC(6:0/6:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40NO8P
CAS Number:
Molecular Weight:
453.51
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99%

form

solution

concentration

20 mg/mL in chloroform

functional group

phospholipid

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCC)=O)COC(CCCCC)=O)=O

InChI

1S/C20H41NO8P/c1-6-8-10-12-19(22)26-16-18(29-20(23)13-11-9-7-2)17-28-30(24,25)27-15-14-21(3,4)5/h18H,6-17H2,1-5H3,(H,24,25)

InChI key

KGWWHBYICAIILE-UHFFFAOYSA-N

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew Hung et al.
Journal of molecular graphics & modelling, 29(5), 597-607 (2010-12-15)
The amyloidogenic peptide apolipoprotein C-II(60-70) is known to exhibit lipid-dependent aggregation behaviour. While the peptide rapidly forms amyloid fibrils in solution, fibrillization is completely inhibited in the presence of lipids. In order to obtain molecular-level insights into the mechanism of
Shuwen Cao et al.
Frontiers in pharmacology, 11, 539-539 (2020-05-20)
The research of lipid nanoparticles (LNPs) has been ongoing for more than three decades, and more research are still being carried out today. Being the first Food and Drug Administration (FDA)-approved nanomedicine, LNPs not only provide various advantages, but also
Andrew Hung et al.
The journal of physical chemistry. B, 113(28), 9447-9453 (2009-06-23)
We investigated the effect of submicellar lipids on amyloid fibril formation. Thioflavin T fluorescence studies showed that submicellar levels of the short-chain phospholipids, dipentanoylphosphatidylcholine and dihexanoylphosphatidylcholine, strongly inhibited amyloid fibril formation by an 11-residue peptide derived from human apolipoprotein C-II
Shogo Taguchi et al.
Colloids and surfaces. B, Biointerfaces, 175, 403-408 (2018-12-18)
The J-aggregate of chlorophyll a (Chla) functions as a light-harvesting antenna in natural systems. In this study, we employed the phospholipid membranes composed of longer-chain 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) and shorter-chain 1,2-dihexanoyl-sn-glycero-3-phosphocholine (DHPC), as a platform to induce Chla aggregates. The DMPC/DHPC
Ronald Soong et al.
Biophysical journal, 97(3), 796-805 (2009-08-05)
Diffusion of various poly(ethylene glycol) (PEG) tracers of well-defined molecular weight and narrow polydispersity confined within the aqueous interstices between positively magnetically aligned bicelles was measured using pulsed-field-gradient (1)H nuclear magnetic resonance. The bicelles consisted of mixtures of dimyristoylphosphatidylcholine (DMPC)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service