Skip to Content
Merck
All Photos(1)

Key Documents

P3690

Sigma-Aldrich

Polyoxyethylene (100) stearate

Synonym(s):

Myrj 59

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900

description

non-ionic

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mona M A Abdel-Mottaleb et al.
International journal of pharmaceutics, 390(2), 208-213 (2010-02-13)
Lipid nanocapsules are recently developed lipid nanocarriers for delivery of lipophilic drugs. Due to their small size and biocompatible nature, lipid nanocapsules (LNC) may be promising carriers for drug delivery with different routes of administration. The aim of this work
Mona M A Abdel-Mottaleb et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 79(1), 36-42 (2011-05-12)
Lipid nanocapsules (LNC) are colloidal carriers providing controlled release profiles and improved bioavailability for many drug substances and diverse administration routes. However, they have not been explored before for transdermal application. Here, we study the behavior of LNC as a
Jiangxiu Niu et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 45(1-2), 216-226 (2011-11-29)
A multifunctional mixed micelle was assembled for drug targeting delivery by combining two newly synthesized amphiphilic polymers, which were octreotide-polyethylene glycol-monostearate (OPMS) and N-octyl-N-succinyl-O-carboxymethyl chitosan (OSCC), respectively. The mixed micelle was designed to be characterized with long circulation, somatostatin receptors
Zhigui Su et al.
Molecular pharmaceutics, 8(5), 1641-1651 (2011-07-21)
A new conjugate, octreotide-polyethylene glycol(100) monostearate (OPMS), was developed for the enhancement of targeting delivery of hydroxycamptothecine (HCPT) loaded in nanostructured lipid carrier (NLC). 2 × 10(-3) and 5 × 10(-3) mmol of OPMS were respectively used to modify NLC
Adam J Shuhendler et al.
Journal of controlled release : official journal of the Controlled Release Society, 157(3), 478-484 (2011-09-24)
For effective and safe thermotherapy, real-time, accurate, three-dimensional tissue thermometry is required. Magnetic resonance imaging (MRI)-based thermometry in combination with current temperature responsive contrast agents only provides an 'off-on' signal at a certain temperature, not indicating temperature increases beyond the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service