Skip to Content
Merck
All Photos(2)

Key Documents

P352-05

Sigma-Aldrich

Porcine Pulmonary Artery Smooth Muscle Cells: PPASMC (Cryovial)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81
Pricing and availability is not currently available.

biological source

Porcine pulmonary artery

Quality Level

form

solid

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 38

morphology

smooth muscle

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cardiovascular diseases

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.


Porcine Pulmonary Artery Smooth Muscle Cells (PPASMC) provide a useful in vitro model to study cellular and molecular mechanisms of pulmonary hypertension characterized by hyperproliferation of pulmonary artery smooth muscle cells.Characterization: positive for smooth muscle cell specific alpha-actin expression.

Cell Line Origin

Artery

Application

cell growth, signaling, receptor transactivation and suppression, vascular remodeling

Components

Porcine Smooth Muscle Cell Basal Medium that contains 10% FBS and 10% DMSO

Preparation Note

  • 1st passage, >500,000 cells in Porcine Smooth Muscle Cell Basal Medium that contains 10% FBS and 10% DMSO
  • Can be cultured at least 16 doublings

Subculture Routine

Please refer to the PPASMC Culture Protocol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service