Skip to Content
Merck
All Photos(1)

Key Documents

P3493

Sigma-Aldrich

Monoclonal Anti-Placenta Growth Factor-2 antibody produced in rat

clone 62526, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-PlGF-2

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€943.00

€943.00


Estimated to ship on29 April 2025


Request a Bulk Order

Select a Size

Change View
500 μG
€943.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

€943.00


Estimated to ship on29 April 2025


Request a Bulk Order

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

62526, monoclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

indirect ELISA: 0.5-1.0 μg/mL
neutralization: suitable
western blot: 1-2 μg/mL

isotype

IgG2a

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Pgf(18654)

General description

The antibody neutralizes receptor-ligand interaction. The antibody shows no cross-reactivity with recombinant human PlGF and recombinant human VEGF.

Immunogen

purified recombinant mouse placenta growth factor 2 expressed in insect Sf 21 cells.

Biochem/physiol Actions

Placental growth factors (PlGF) are involved in the growth, migration and survival of endothelial cells. All the four isoforms identified are expressed in placenta and umbilical vein endothelial cells. PlGF bind to specific Flt-1 receptors that result in activation and recruitment of dendritic cells, smooth muscle cells and inflammatory cells. PlGF2 has the ability to bind heparin while PlGF1 is a soluble isoform.

Physical form

Lyophilized from a 0.2 μm filtered solution of phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valeria Tarallo et al.
American journal of cancer research, 1(2), 265-274 (2011-10-05)
Angiogenesis is one of the crucial events for cancer development and growth and vascular endothelial growth factor (VEGF) family plays an essential role in this biological phenomenon. The members of VEGF family mainly involved in angiogenesis are VEGF-A, VEGF-B and
S C W Tang et al.
Kidney international, 73(3), 288-299 (2007-11-23)
Angiotensin-converting enzyme inhibitors, angiotensin II receptor blockers, and statins have renoprotective effects. We studied the cellular mechanisms for this effect in adriamycin-treated mice receiving captopril, losartan, simvastatin, or their combinations. The mice developed albuminuria, renal insufficiency, and parenchymal inflammation/fibrosis accompanied

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service