Recommended Products
form
powder
Quality Level
mp
158-162 °C (lit.)
storage temp.
−20°C
SMILES string
Cl.COC(=O)[C@@H](N)Cc1ccccc1
InChI
1S/C10H13NO2.ClH/c1-13-10(12)9(11)7-8-5-3-2-4-6-8;/h2-6,9H,7,11H2,1H3;1H/t9-;/m0./s1
InChI key
SWVMLNPDTIFDDY-FVGYRXGTSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Immunopharmacology, 28(1), 39-45 (1994-07-01)
Monocytes macrophages have negative regulatory effects on many immunologic responses. Depletion of monocytes from peripheral blood using the lysosomotropic agent, L-phenylalanine methyl ester (PME), has been shown to improve lymphokine activated killer (LAK) cell expansion in vitro. A pilot study
Biochimica et biophysica acta, 1758(3), 347-354 (2005-12-29)
Many promising therapeutics are currently awaiting their clinical application. Due to their low capability of cell membrane crossing, these compounds do not reach their site of action. One way to overcome this problem might be the fusion of these agents
Chemical communications (Cambridge, England), (1)(1), 134-135 (2003-03-04)
Reaction of ADP with amino acid methyl esters mediated by trimethylsilyl chloride in pyridine produced adenosine 5'-phosphoramidates in good yields under mild conditions, it is interesting that nucleophilic attack of amino acid methyl esters only occurred on alpha-phosphorus of ADP.
Biotechnology progress, 27(6), 1554-1560 (2012-01-12)
The direct one-step synthesis of L-phenylalanine methyl ester in an organic-aqueous biphasic system using phenylalanine ammonia lyase (E.C.4.3.1.5, PAL) containing Rhodotorula glutinis yeast whole cells was reported earlier. We report here further optimization of this biotransformation using isolated PAL, when
Organic & biomolecular chemistry, 3(8), 1435-1447 (2005-04-14)
The highly diastereoselective samarium diiodide and D(2)O-promoted conjugate reduction of homochiral (E)- and (Z)-benzylidene and isobutylidene diketopiperazines (E)-5,7 and (Z)-6,8 has been demonstrated. This methodology allows the asymmetric synthesis of methyl (2S,3R)-dideuteriophenylalanine 27 in > or = 95% de and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service