Skip to Content
Merck

Skip To

P2334

Phenyl-Sepharose 6 Fast Flow

low substitution, extent of labeling: ~20 μmol per mL

Synonym(s):

Phenyl-Agarose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mL
Check Cart for Availability
€ 370,00

About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
MDL number:

€ 370,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

suspension

extent of labeling

~20 μmol per mL

matrix

agarose, 6% crosslinked

matrix activation

epichlorohydrin

matrix attachment

ether

matrix spacer

3 atoms

particle size

45-165 μm

capacity

~14 mg/mL binding capacity (BSA)(lineal flow rate of 100 cm/hr)

storage temp.

2-8°C

General description

P2334-200ml′s updated product number is GE17-0965-05

Application

Phenyl [YM="Sepharose"] is used in protein chromatography, affinity chromatography, hydrophobic interaction media, resins and separation media. Phenyl Sepharose has been used in studies that contributed to improving industrial applications in additives in detergents and feed industries. Phenyl sepharose has also been used to study microbial communities inhabiting hypersaline environments.

Physical form

Suspension in 20% ethanol
aqueous ethanol suspension

Legal Information

Sepharose is a trademark of Cytiva

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P2459P78926B100
particle size

45-165 μm

particle size

45-165 μm

particle size

45-165 μm

particle size

45-165 μm (wet)

matrix

agarose, 6% crosslinked

matrix

agarose, 6% cross-linked

matrix

agarose, 4% cross-linked

matrix

6% agarose

form

suspension

form

suspension

form

suspension

form

suspension

capacity

~14 mg/mL binding capacity (BSA)(lineal flow rate of 100 cm/hr)

capacity

-

capacity

15-20 mg/mL binding capacity (human serum albumin), 3-5 mg/mL binding capacity (β-lactoglobulin)

capacity

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

-


Still not finding the right product?

Explore all of our products under Phenyl-Sepharose 6 Fast Flow


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

95.0 °F

flash_point_c

35 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters