Skip to Content
Merck
All Photos(1)

Documents

P2032

Sigma-Aldrich

Pepstatin A−Agarose

saline suspension

Synonym(s):

Pepstatin A resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

biological source

microbial (fermentation)
plant

form

saline suspension

technique(s)

affinity chromatography: suitable

matrix

cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

carboxyl

matrix spacer

9 atoms

capacity

20-40 mg/mL binding capacity (pepsin)

suitability

suitable for chromatography

storage temp.

2-8°C

Related Categories

Application

Pepstatin A-agarose is used in protein chromatography, affinity chromatography and specialty resins. Pepstatin A-agarose has been used to characterize three chitosanase isozymes isolated from a commercial crude porcine pepsin preparation.

Physical form

Suspension in 0.5 M NaCl containing preservative

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madanan Madathiparambil Gopalakrishnan et al.
The Biochemical journal, 383(Pt. 3), 507-515 (2004-07-17)
Before delivery to endosomes, portions of proCD (procathepsin D) and proSAP (prosaposin) are assembled into complexes. We demonstrate that such complexes are also present in secretions of cultured cells. To study the formation and properties of the complexes, we purified
F Canduri et al.
Biochemistry and molecular biology international, 45(4), 797-803 (1998-08-26)
Cathepsin D, a lysosomal aspartic protease, has been purified from porcine liver using a combination of pepstatin-A agarose and Affi-Gel Blue affinity chromatography, followed by size-exclusion chromatography. The purified protein consists of two polypeptide chains of 15 and 30 kDa
Mark S Pearson et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(9), 3007-3019 (2009-04-22)
Hookworms digest hemoglobin from erythrocytes via a proteolytic cascade that begins with the aspartic protease, APR-1. Ac-APR-1 from the dog hookworm, Ancylostoma caninum, protects dogs against hookworm infection via antibodies that neutralize enzymatic activity and interrupt blood-feeding. Toward developing a
H S Kim et al.
Journal of immunology (Baltimore, Md. : 1950), 165(6), 3268-3274 (2000-09-07)
The intestinal epithelium forms a first line of innate host defense by secretion of proteins with antimicrobial activity against microbial infection. Despite the extensive studies on the antimicrobial host defense in many gastrointestinal tracts, little is known about the antimicrobial
Jonathan A Green et al.
Theriogenology, 63(5), 1481-1503 (2005-02-24)
The pregnancy-associated glycoproteins (PAGs) are a large gene family expressed in trophoblast cells of ruminant ungulates. The detection of PAGs (more specifically, PAG-1) in maternal serum has served as the basis for pregnancy detection in cattle. Unfortunately, PAG-1 and/or antigenically-related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service