The recommended storage temperature is -20 °C.
Product Name
Pituitary Extract bovine, solution, ~14 mg/mL protein in PBS, 0.2 μm filtered
biological source
bovine
sterility
0.2 μm filtered
form
solution
quality
endotoxin tested
packaging
pkg of 2.5 and 25 mL
concentration
~14 mg/mL protein in PBS
impurities
<50 EU/mL
color
brown
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Pituitary Extract, bovine (PEX) is an extract of bovine pituitary tissue containing growth factors. PEX can be used to promote the growth of several types of human epithelial cells in culture. Pituitary extract, bovine, is prepared using a modification of a published procedure.
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J C Willey et al.
Journal of cellular physiology, 124(2), 207-212 (1985-08-01)
The mechanisms of action of extracellular mitogens for normal human bronchial epithelial cells (NHBE) were investigated by observing their effects on selected biochemical pathways when the cells were incubated in serum-free media. We find that (a) epidermal growth factor (EGF)
Jie Liu et al.
Animal reproduction science, 115(1-4), 49-57 (2008-12-26)
Different culture systems were evaluated for their ability to support attachment and proliferation of the somatic cells obtained from ovine semen. Ejaculates (n=14) were collected from eight rams representing three breeds, Dorper, Suffolk and Hampshire. All samples were processed immediately
S L Hammond et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(17), 5435-5439 (1984-09-01)
A serum-free medium with bovine pituitary extract as the only undefined supplement has been developed for long-term culture of human mammary epithelial cells. This medium supports serial subculture of normal cells for 10-20 passages (1:10 splits) without conditioning or special
Li Na Zhou et al.
Stem cell research & therapy, 11(1), 153-153 (2020-04-11)
To date, it has repeatedly been demonstrated that infusing bone marrow-derived stem cells (BMSCs) into acellular nerve scaffolds can promote and support axon regeneration through a peripheral nerve defect. However, harvesting BMSCs is an invasive and painful process fraught with
Yan Hu et al.
Stem cells (Dayton, Ohio), 38(11), 1467-1478 (2020-06-12)
Wnt/β-catenin signaling regulates progenitor cell fate decisions during lung development and in various adult tissues. Ectopic activation of Wnt/β-catenin signaling promotes tissue repair in emphysema, a devastating lung disease with progressive loss of parenchymal lung tissue. The identity of Wnt/β-catenin
-
What are the storage conditions for this product?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service