Skip to Content
Merck
All Photos(3)

Key Documents

P0749

Sigma-Aldrich

Anti-Potassium Channel Kv11.1 Extracellular antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-HERG, Anti-KCNH2

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€759.00

€759.00


Estimated to ship on29 May 2025



Select a Size

Change View
200 μL
€759.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€759.00


Estimated to ship on29 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

flow cytometry: 2-8 μg using live intact K562 (human chronic myelogenous leukemia) cells (per 106 cells)
immunocytochemistry: 1:25
western blot: 1:200 using HEK-KV11.1 transfected cells

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNH2(3757)
mouse ... Kcnh2(16511)
rat ... Kcnh2(117018)

General description

The K+ channel family has been sub-grouped in to 2 TM, 4 TM and 6 TM K+ channels based on the number of transmembrane domains. The voltage-gated Potassium or Kv channels belong to the 6 TM K+ channel family that also include other sub-types such as the hERG, KCNQ, and EAG channels. Kv11.1 belongs to the EAG family and facilitates the repolarization of normal action potential in heart muscles. Genetic alterations in Kv11.1 can cause heart disorders such as long QT syndrome, sudden cardiac death and arhythmias. Kv11.1 also regulates cell growth and hence may be studied as a target for cancer treatment. Anti-Potassium channel Kv11.1 extracellular antibodies are specific to Kv11.1.

Immunogen

peptide AFLLKETEEGPPATEC corresponding to residues 430-445 of human KV11.1. This sequence has 11/16 residues identical in mouse and rat and has 15/16 residues identical in rabbit and dog.

Application

Anti-Potassium channel Kv11.1 extracellular antibodies may be used in immunofluorescence microscopy . It may also be used for the detection of Kv11.1 by techniques such as immunocytochemistry, flow cytometry using live intact K562 (human chronic myelogenous leukemia) cells and western blotting.

Physical form

Lyophilized powder from phosphate buffered saline containing 1% bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cell Surface Expression of Human Ether-a-go-go-Related Gene (hERG) Channels is Regulated by Caveolin-3 via the Ubiquitin Ligase Nedd4-2
Guo, J.
The Journal of Biological Chemistry, (287(40), 33132-33141 (2012)
Jingyun Dong et al.
Heliyon, 4(12), e01015-e01015 (2018-12-26)
Molecular testing of the deceased (Molecular Autopsy) is an overlooked area in the United States healthcare system and is not covered by medical insurance, leading to ineffective care for surviving families of thousands of sudden unexpected natural deaths each year.
Audrey Farrugia et al.
International journal of molecular sciences, 22(17) (2021-09-11)
Long QT syndrome is one of the most common hereditary channelopathies inducing fatal arrhythmias and sudden cardiac death. We identified in a sudden arrhythmic death syndrome case a C-term KCNH2 mutation (c.3457C > T; p.His1153Tyr) classified as variant of unknown

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service