Skip to Content
Merck

Skip To

P0671

Poly-DL-ornithine hydrobromide

suitable for cell culture, Mol wt >30,000

Synonym(s):

DL-Ornithine homopolymer hydrobromide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Poly-DL-ornithine hydrobromide, mol wt >30,000

form

powder or solid

Quality Segment

mol wt

>30,000

technique(s)

cell culture | mammalian: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

Br.NCCCC(N)C(O)=O

InChI

1S/C5H12N2O2.BrH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H

InChI key

GWRQMKDBBHFVIZ-UHFFFAOYSA-N

Biochem/physiol Actions

Poly-DL-ornithine hydrobromide has applications as a surface complex.

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P8638P3655P4538
form

powder or solid

form

powder or solid

form

powder

form

powder

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

color

white to off-white

color

white to off-white

color

white to off-white

color

white to off-white

mol wt

>30,000

mol wt

3,000-15,000

mol wt

30,000-70,000

mol wt

5,000-15,000


Still not finding the right product?

Explore all of our products under Poly-DL-ornithine hydrobromide


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters